Anti-JUNB Goat Antibody

Supplier: OriGene
ORIGTA305921EA 456 EUR
ORIGTA305921
Anti-JUNB Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: JUNB (jun B proto-oncogene)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR