Anti-HCAR1 Goat Antibody

Supplier: OriGene
ORIGTA305922EA 547 EUR
ORIGTA305922
Anti-HCAR1 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 03-1?g/ml

Type: Primary
Antigen: HCAR1 (hydroxycarboxylic acid receptor 1)
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR