Anti-PKD2 Goat Antibody

Supplier: OriGene
ORIGTA305931EA 454 EUR
ORIGTA305931
Anti-PKD2 Goat Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-3?g/ml

Type: Primary
Antigen: PKD2 (polycystic kidney disease 2 (autosomal dominant))
Clonality:
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG (Goat)
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR