Anti-CXCR4 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA305935EA 600 EUR
ORIGTA305935
Anti-CXCR4 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunochemistry: 10 ?g/ml; Immunoprecipitation: 10 ?g/ml; Immunofluorescence: 20 ?g/ml; Flow Cytometry: 20 ?g/ml

Type: Primary
Antigen: CXCR4 (chemokine (C-X-C motif) receptor 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR