Anti-CCR5 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA305941EA 600 EUR
ORIGTA305941
Anti-CCR5 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Immunochemistry: 20 ?g/ml

Type: Primary
Antigen: CCR5 (chemokine (C-C motif) receptor 5 (gene/pseudogene))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR