Anti-NRTN Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA305946EA 494 EUR
ORIGTA305946
Anti-NRTN Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 5?g/ml

Type: Primary
Antigen: NRTN (neurturin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR