Anti-GFRA3 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA305954EA 595 EUR
ORIGTA305954
Anti-GFRA3 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: GFRA3 (GDNF family receptor alpha 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR