Anti-TNFRSF10A Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA305955EA 549 EUR
ORIGTA305955
Anti-TNFRSF10A Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.5 mg/ml; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: TNFRSF10A (tumor necrosis factor receptor superfamily, member 10a)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR