Anti-CFLAR Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA305960EA 600 EUR
ORIGTA305960
Anti-CFLAR Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.5 ?g/ml; Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: CFLAR (CASP8 and FADD-like apoptosis regulator)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR