Anti-CXCR6 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA305967EA 499 EUR
ORIGTA305967
Anti-CXCR6 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: CXCR6 (chemokine (C-X-C motif) receptor 6)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR