Anti-TNFRSF10B Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA305974EA 499 EUR
ORIGTA305974
Anti-TNFRSF10B Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 2 ?g/ml; Immunochemistry: 5 ?g/ml; Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: TNFRSF10B (tumor necrosis factor receptor superfamily, member 10b)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR