Anti-TNFRSF10D Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA305975EA 549 EUR
ORIGTA305975
Anti-TNFRSF10D Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:1000 Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: TNFRSF10D (tumor necrosis factor receptor superfamily, member 10d, decoy with truncated death domain)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR