Anti-ADAM10 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA305977EA 549 EUR
ORIGTA305977
Anti-ADAM10 Rabbit Polyclonal Antibody
Antibodies
  • Recommended Dilutions: Western Blot: 1:1000-1:2000 Immunochemistry; Immunofluorescence: 10 μg/ml

Type: Primary
Antigen: ADAM10 (ADAM metallopeptidase domain 10)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR