Anti-DAPK3 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA305981EA 499 EUR
ORIGTA305981
Anti-DAPK3 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunochemistry: 10 ?g/ml; Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: DAPK3 (death-associated protein kinase 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR