Anti-CASP9 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA305982EA 549 EUR
ORIGTA305982
Anti-CASP9 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: CASP9 (caspase 9, apoptosis-related cysteine peptidase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR