Anti-CCR8 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA305990EA 499 EUR
ORIGTA305990
Anti-CCR8 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: CCR8 (chemokine (C-C motif) receptor 8)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR