Anti-DFFB Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA305993EA 600 EUR
ORIGTA305993
Anti-DFFB Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot Immunochemistry: 5 ?g/ml

Type: Primary
Antigen: DFFB (DNA fragmentation factor, 40kDa, beta polypeptide (caspase-activated DNase))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR