Anti-MYD88 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA305999EA 494 EUR
ORIGTA305999
Anti-MYD88 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: MYD88 (myeloid differentiation primary response 88)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR