Anti-IL1RAP Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306000EA 549 EUR
ORIGTA306000
Anti-IL1RAP Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: IL1RAP (interleukin 1 receptor accessory protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR