Anti-STK17B Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306007EA 494 EUR
ORIGTA306007
Anti-STK17B Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: STK17B (serine/threonine kinase 17b)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR