Anti-NOL3 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306014EA 499 EUR
ORIGTA306014
Anti-NOL3 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1:500

Type: Primary
Antigen: NOL3 (nucleolar protein 3 (apoptosis repressor with CARD domain))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR