Anti-ACIN1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306019EA 549 EUR
ORIGTA306019
Anti-ACIN1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.5 ?g/ml; Immunochemistry: 0.5 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: ACIN1 (apoptotic chromatin condensation inducer 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR