Anti-TNFSF13 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306022EA 600 EUR
ORIGTA306022
Anti-TNFSF13 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 4-5 ?g/ml

Type: Primary
Antigen: TNFSF13 (tumor necrosis factor (ligand) superfamily, member 13)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR