Anti-AIFM1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306028EA 549 EUR
ORIGTA306028
Anti-AIFM1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunochemistry: 2 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: AIFM1 (apoptosis-inducing factor, mitochondrion-associated, 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR