Anti-FAIM Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306048EA 544 EUR
ORIGTA306048
Anti-FAIM Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 5-10 ?g/ml

Type: Primary
Antigen: FAIM (Fas apoptotic inhibitory molecule)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR