Anti-TNFRSF17 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306065EA 499 EUR
ORIGTA306065
Anti-TNFRSF17 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 2-5 ?g/ml

Type: Primary
Antigen: TNFRSF17 (tumor necrosis factor receptor superfamily, member 17)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR