Anti-PCBP4 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306079EA 600 EUR
ORIGTA306079
Anti-PCBP4 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.5-2 ?g/ml; Immunochemistry: 1 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: PCBP4 (poly(rC) binding protein 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR