Anti-CARD9 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306093EA 600 EUR
ORIGTA306093
Anti-CARD9 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 2.5 ?g/ml; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: CARD9 (caspase recruitment domain family, member 9)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR