Anti-KLHL1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306098EA 494 EUR
ORIGTA306098
Anti-KLHL1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 2-4?g/ml; Immunochemistry: 10?g/ml

Type: Primary
Antigen: KLHL1 (kelch-like family member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR