Anti-BMF Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306103EA 595 EUR
ORIGTA306103
Anti-BMF Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 2 ?g/ml

Type: Primary
Antigen: BMF (Bcl2 modifying factor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR