Anti-NLRP1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306106EA 600 EUR
ORIGTA306106
Anti-NLRP1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-4 ?g/ml; Immunochemistry: 10 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: NLRP1 (NLR family, pyrin domain containing 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR