Anti-BBC3 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306108EA 499 EUR
ORIGTA306108
Anti-BBC3 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 2 ?g/ml; Immunochemistry; Immunofluorescence: 1 ?g/ml

Type: Primary
Antigen: BBC3 (BCL2 binding component 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR