Anti-ANP32E Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306128EA 499 EUR
ORIGTA306128
Anti-ANP32E Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.5-2 ?g/ml

Type: Primary
Antigen: ANP32E (acidic (leucine-rich) nuclear phosphoprotein 32 family, member E)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR