Anti-ACE2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306148EA 494 EUR
ORIGTA306148
Anti-ACE2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.5-2 ?g/ml

Type: Primary
Antigen: ACE2 (angiotensin I converting enzyme 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR