Anti-TRIM5 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306152EA 499 EUR
ORIGTA306152
Anti-TRIM5 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: TRIM5 (tripartite motif containing 5)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR