Anti-SIGIRR Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306181EA 499 EUR
ORIGTA306181
Anti-SIGIRR Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.5-2 ?g/ml; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: SIGIRR (single immunoglobulin and toll-interleukin 1 receptor (TIR) domain)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR