Anti-CASP1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306199EA 549 EUR
ORIGTA306199
Anti-CASP1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.5 ?g/ml

Type: Primary
Antigen: CASP1 (caspase 1, apoptosis-related cysteine peptidase)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR