Anti-RPS6KA1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306221EA 600 EUR
ORIGTA306221
Anti-RPS6KA1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 2.5-10 ?g/ml; Immunochemistry: 10 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: RPS6KA1 (ribosomal protein S6 kinase, 90kDa, polypeptide 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR