Anti-C1QTNF4 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306235EA 499 EUR
ORIGTA306235
Anti-C1QTNF4 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-4 ?g/ml; Immunochemistry: 10 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: C1QTNF4 (C1q and tumor necrosis factor related protein 4)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR