Anti-NGFR Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306246EA 600 EUR
ORIGTA306246
Anti-NGFR Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1 ?g/ml; Immunochemistry: 10 ?g/ml

Type: Primary
Antigen: NGFR (nerve growth factor receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR