Anti-IL32 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306297EA 499 EUR
ORIGTA306297
Anti-IL32 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 2.5-5 ?g/ml

Type: Primary
Antigen: IL32 (interleukin 32)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR