Anti-BMI1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306298EA 499 EUR
ORIGTA306298
Anti-BMI1 Rabbit Polyclonal Antibody
Antibodies
  • Recommended Dilutions: Western Blot: 0.5-2 μg/ml; Immunochemistry: 10 μg/ml; Immunofluorescence: 20 μg/ml

Type: Primary
Antigen: BMI1 (BMI1 polycomb ring finger oncogene)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR