Anti-SNAI2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306361EA 499 EUR
ORIGTA306361
Anti-SNAI2 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.5-2 ?g/ml; Immunofluorescence

Type: Primary
Antigen: SNAI2 (snail family zinc finger 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR