Anti-DARC Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306406EA 499 EUR
ORIGTA306406
Anti-DARC Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: DARC (Duffy blood group, atypical chemokine receptor)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR