Anti-NPAS3 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306418EA 499 EUR
ORIGTA306418
Anti-NPAS3 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: NPAS3 (neuronal PAS domain protein 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR