Anti-ORAI3 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306430EA 595 EUR
ORIGTA306430
Anti-ORAI3 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 2-4 ?g/ml; Immunochemistry: 5 ?g/ml; Immunofluorescence: 10 ?g/ml

Type: Primary
Antigen: ORAI3 (ORAI calcium release-activated calcium modulator 3)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR