Anti-TBC1D1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306433EA 600 EUR
ORIGTA306433
Anti-TBC1D1 Rabbit Polyclonal Antibody
Antibodies
  • Recommended Dilutions: Western Blot: 2-4 μg/ml

Type: Primary
Antigen: TBC1D1 (TBC1 (tre-2/USP6, BUB2, cdc16) domain family, member 1)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR