Anti-TBC1D10C Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306446EA 549 EUR
ORIGTA306446
Anti-TBC1D10C Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 1-2 ?g/ml

Type: Primary
Antigen: TBC1D10C (TBC1 domain family, member 10C)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR