Anti-ATG16L1 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306514EA 499 EUR
ORIGTA306514
Anti-ATG16L1 Rabbit Polyclonal Antibody
Antibodies
Recommended Dilutions: Western Blot: 0.25-0.5 ?g/ml; Immunochemistry: 2 ?g/ml; Immunofluorescence: 4 ?g/ml

Type: Primary
Antigen: ATG16L1 (autophagy related 16-like 1 (S. cerevisiae))
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR