Anti-SIRT2 Rabbit Polyclonal Antibody

Supplier: OriGene
ORIGTA306535EA 549 EUR
ORIGTA306535
Anti-SIRT2 Rabbit Polyclonal Antibody
Antibodies
  • Recommended Dilutions: Western Blot: 2.5-5 μg/ml; Immunochemistry: 2.5 μg/ml; Immunofluorescence: 20 μg/ml

Type: Primary
Antigen: SIRT2 (sirtuin 2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR